missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-methoxybenzoic acid, TRC

Product Code. 30396106
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396106 500 mg -
30394839 100 mg 100mg
30403847 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396106 Supplier Toronto Research Chemicals Supplier No. TRCH855035500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61227-25-6
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
IUPAC Name 5-hydroxy-2-methoxybenzoic acid
SMILES COc1ccc(O)cc1C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H8O4/c1-12-7-3-2-5(9)4-6(7)8(10)11/h2-4,9H,1H3,(H,10,11)
Formula Weight 168.0423
Chemical Name or Material 5-Hydroxy-2-methoxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.