missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-iodobenzaldehyde, 97%, Thermo Scientific™

Product Code. 10376714
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10376714 10 g 10g
10316674 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10376714 Supplier Thermo Scientific Maybridge Supplier No. MO07995EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 50765-11-2
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.019
MDL Number MFCD09025923
InChI Key JRJRYFRHGBHRHI-UHFFFAOYSA-N
Synonym 5-hydroxy-2-iodo-benzaldehyde, 2-jod-5-oxy-benzaldehyd, benzaldehyde,5-hydroxy-2-iodo
PubChem CID 279261
IUPAC Name 5-hydroxy-2-iodobenzaldehyde
SMILES C1=CC(=C(C=C1O)C=O)I

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 248.01
Percent Purity 95%
Chemical Name or Material 5-Hydroxy-2-iodobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.