missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-adamantanone, 98%, Thermo Scientific™

Product Code. 10061324 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10061324 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10061324 Supplier Thermo Scientific Acros Supplier No. 426330010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 20098-14-0
CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H14O2
MDL Number MFCD00192211
Quantity 1 g
Synonym idramantone, 5-hydroxy-2-adamantanone, kemantane, 5-hydroxy-adamantan-2-one, kemantan, 5-hydroxytricyclo 3.3.1.1 3,7 decanone, tricyclo 3.3.1.13,7 decanone, 5-hydroxy, idramantone inn, 4-oxo-1-adamantanol
InChI Key TZBDEVBNMSLVKT-UHFFFAOYSA-N
SMILES C1C2CC3CC(C2)(CC1C3=O)O
IUPAC Name 5-hydroxyadamantan-2-one
Molecular Weight (g/mol) 166.22
PubChem CID 64184
ChEBI CHEBI:48581
Formula Weight 166.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxy-2-adamantanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.