missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-1-tetralone, 99%

Product Code. 11498886
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498886 5 g 5g
11488886 1 g 1g
11408896 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498886 Supplier Thermo Scientific Alfa Aesar Supplier No. A17034.06
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28315-93-7 | C10H10O2 | 162.188 g/mol

5-Hydroxy-1-tetralone, is used as a metabolite of Levobunolol (L335000) and d-Bunolol (L335010). It is also used as a reagent for the determination of hexoses and oligosaccharides by a fluorescence technique. A reagent that is used for fluorescence determination of enzyme activity. Some of the other applications include as internal standard during HPLC determination of 4-hydroxymephenytoin (4-OH-M) in human urine, as fluorescent labeling reagent during micro detection of glycosphingolipid on TLC plates, in synthesis of 1,2,3,4-tetrahydro-5H-1-benzazepine-quinone derivatives, in synthesis of new chiral oxathiane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hydroxy-1-tetralone, is used as a metabolite of Levobunolol (L335000) and d-Bunolol (L335010). It is also used as a reagent for the determination of hexoses and oligosaccharides by a fluorescence technique. A reagent that is used for fluorescence determination of enzyme activity. Some of the other applications include as internal standard during HPLC determination of 4-hydroxymephenytoin (4-OH-M) in human urine, as fluorescent labeling reagent during micro detection of glycosphingolipid on TLC plates, in synthesis of 1,2,3,4-tetrahydro-5H-1-benzazepine-quinone derivatives, in synthesis of new chiral oxathiane.

Solubility
Soluble in alcohol, ether, benzene and acetic acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, acid chlorides, acid anhydrides. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28315-93-7
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00001693
InChI Key YPPZCRZRQHFRBH-UHFFFAOYSA-N
Synonym 5-hydroxy-1-tetralone, 1-tetralon-5-ol, 5-hydroxy-3,4-dihydronaphthalen-1 2h-one, 5-hydroy-1-tetraione, 1 2h-naphthalenone, 3,4-dihydro-5-hydroxy, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 5-hydroxytetralone, 3,4-dihydro-5-hydroxy-1 2h-naphthalenone, 1,2,3,4-tetrahydro-5-hydroxynaphthalen-1-one, 5-hydroxy-2,3,4-trihydronaphthalen-1-one
PubChem CID 119921
IUPAC Name 5-hydroxy-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=CC=C2O)C(=O)C1

Specifications

Melting Point 209°C to 211°C
Quantity 5 g
Beilstein 2437410
Solubility Information Soluble in alcohol,ether,benzene and acetic acid.
Formula Weight 162.19
Percent Purity 99%
Chemical Name or Material 5-Hydroxy-1-tetralone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.