missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-1,4-naphthoquinone, TRC

Product Code. 30395610
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395610 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395610 Supplier Toronto Research Chemicals Supplier No. TRCH9495602.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Juglone
CAS 481-39-0
Molecular Formula C10 H6 O3
Quantity 2.5 g
Synonym 1,4-Naphthalenedione, 5-hydroxy-, 1,4-Naphthoquinone, 5-hydroxy- (8CI), Juglone (6CI), 1,4-Dihydro-1,4-dioxo-5-hydroxynaphthalene, 5-Hydroxy-1,4-naphthalenedione, 5-Hydroxy-1,4-naphthoquinone, 5-Hydroxynaphthoquinone, 8-Hydroxy-1,4-naphthoquinone, Akhnot, C.I. 75500, C.I. Natural Brown 7, Juglon, NSC 153189, NSC 34266, NSC 622948, Natural Brown 7, Nucin, Regianin, Walnut Extract
InChI Formula InChI=1S/C10H6O3/c11-7-4-5-9(13)10-6(7)2-1-3-8(10)12/h1-5,12H
SMILES Oc1cccc2C(=O)C=CC(=O)c12
IUPAC Name 5-hydroxynaphthalene-1,4-dione
Molecular Weight (g/mol) 174.15
Formula Weight 174.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.