missing translation for 'onlineSavingsMsg'
Learn More

5-HT7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

£153.00 - £406.00

Specifications

Antigen 5-HT7
Concentration LYOPH
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18206281
View Documents
Novus Biologicals
NBP1-46598-0.025ML
0.025 mL
£153.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18735313
View Documents
Novus Biologicals
NBP1-46598
0.1 mL
£406.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

5-HT7 Polyclonal specifically detects 5-HT7 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

5-HT7
Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot
Unconjugated
Rabbit
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
P34969
3363
IgG
Unpurified
Specific for 5HT7R receptor.
LYOPH
Polyclonal
Antisera
RUO
Mouse, Guinea Pig, Rat, Human
5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), serotonin 5-HT-7 receptor, Serotonin receptor 7
HTR7
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.