missing translation for 'onlineSavingsMsg'
Learn More

5-HT7 Antibody (1015322) [Janelia Fluor™ 635], Novus Biologicals™

Product Code. 30167163 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30167163 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30167163 Supplier Novus Biologicals Supplier No. FAB10325JF635

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

5-HT7 Monoclonal antibody specifically detects 5-HT7 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5-HT7
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 1015322
Conjugate Janelia Fluor 635
Formulation 50mM Sodium Borate
Gene Alias 5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), serotonin 5-HT-7 receptor, Serotonin receptor 7
Host Species Mouse
Immunogen Synthetic peptide containing human 5-HTR7, Accession # P34969
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 3363
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.