missing translation for 'onlineSavingsMsg'
Learn More

5-HT4 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18406007 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406007 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406007 Supplier Novus Biologicals Supplier No. NBP178403JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

5-HT4 Polyclonal antibody specifically detects 5-HT4 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5-HT4
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias 5-HT-4, 5-HT4R, 5-hydroxytryptamine (serotonin) receptor 4,5-hydroxytryptamine receptor 4,5-HT45-hydroxytryptamine4 receptor, cardiac 5-HT4 receptor, Serotonin receptor 4
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human 5HT4 Receptor protein (between residues 350-388) [UniProt Q13639, isoform 1]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, GPCR, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 3360
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.