missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT4 Antibody (631769), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6545P
This item is not returnable.
View return policy
Description
5-HT4 Monoclonal antibody specifically detects 5-HT4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifications
| 5-HT4 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry | |
| 631769 | |
| Flow Cytometry, Immunohistochemistry | |
| 5-HT-4, 5-HT4R, 5-hydroxytryptamine (serotonin) receptor 4,5-hydroxytryptamine receptor 4,5-HT45-hydroxytryptamine4 receptor, cardiac 5-HT4 receptor, Serotonin receptor 4 | |
| NS0 mouse myeloma cell line transfected with human 5-HT4 | |
| 0.1 mL | |
| Asthma, GPCR, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 3360 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction