missing translation for 'onlineSavingsMsg'
Learn More

5-HT3E Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18332000 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332000 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18332000 Supplier Novus Biologicals Supplier No. NBP2825410.1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

5-HT3E Polyclonal specifically detects 5-HT3E in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5-HT3E
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias 5-HT3 receptor subunit E splice variant HTR3Ea, 5-HT3c1,5-HT3E, 5-hydroxytryptamine (serotonin) receptor 3, family member E, 5-hydroxytryptamine receptor 3E, MGC120035, MGC120036, serotonin receptor 3E
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of Human 5-HT3E
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285242
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.