missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT3A Antibody (3B11C10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£179.00 - £477.00
Specifications
| Antigen | 5-HT3A |
|---|---|
| Clone | 3B11C10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18055686
|
Novus Biologicals
NBP2-61781 |
0.1 mL |
£477.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657496
|
Novus Biologicals
NBP2-61781-0.025ml |
0.025 mL |
£179.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-HT3A Monoclonal specifically detects 5-HT3A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| 5-HT3A | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 3359 | |
| Purified recombinant fragment of human HTR3A (AA: extra 24-157) expressed in E. Coli. | |
| Primary | |
| 55.3 kDa |
| 3B11C10 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| 5-HT-3,5-HT3-A, 5-HT3R5-hydroxytryptamine receptor 3A, 5-hydroxytryptamine (serotonin) receptor 3A, 5-hydroxytryptamine receptor 3,5-HT3A5HT3R, HTR35HT3 serotonin receptor, Serotonin receptor 3A, Serotonin-gated ion channel receptor | |
| HTR3A | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title