missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ 5-HT2B Antibody (CL14576), Novus Biologicals™
Mouse Monoclonal Antibody
£336.00 - £483.00
Specifications
| Antigen | 5-HT2B |
|---|---|
| Clone | CL14576 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043034
|
Novus Biologicals™
NBP3-26870-25ul |
25 μL |
£336.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043792
|
Novus Biologicals™
NBP3-26870-100ul |
100 μL |
£483.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-HT2B Monoclonal antibody specifically detects 5-HT2B in Human samples. It is validated for Western BlotSpecifications
| 5-HT2B | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS, pH 7.2, 40% glycerol | |
| 3357 | |
| IgG1 | |
| Protein A purified |
| CL14576 | |
| Western Blot 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 5-HT(2B), 5-HT-2B, 5-HT2B5-HT 2B receptor, 5-hydroxytryptamine (serotonin) receptor 2B, 5-hydroxytryptamine 2B receptor, 5-hydroxytryptamine receptor 2B, Serotonin receptor 2B | |
| This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of P41595. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title