missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
5-HT1A Polyclonal antibody specifically detects 5-HT1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | 5-HT1A |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | 5-HT-1A, 5-HT1a receptor, 5-HT1A5-hydroxytryptamine receptor 1A, 5-hydroxytryptamine (serotonin) receptor 1A, ADRB2RL15HT1a, ADRBRL1, G protein coupled receptor, G-21, guanine nucleotide-binding regulatory protein-coupled receptor, Serotonin receptor 1A |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: VEKTGADTRHGASPAPQPKKSVNGESGSRNWRLGVESKAGGALCANGAVRQGDDGAALEVIEVHRVGNSKEHLPLPSEAGPTPCAPASFERKNERNAEAKRKMALA |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?