missing translation for 'onlineSavingsMsg'
Learn More

5-Hexynoic acid, 97%

Product Code. 10210403 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10210403 1 g Glass bottle 1g
10371023 5 g Glass bottle 5g
10686894 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10210403 Supplier Thermo Scientific Acros Supplier No. 383700010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53293-00-8 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53293-00-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00066346
InChI Key VPFMEXRVUOPYRG-UHFFFAOYSA-N
Synonym 5-hexynoic acid, 5-hexyn-1-oic acid, 4-ethynylbutyric acid, acmc-1an4h
PubChem CID 143036
ChEBI CHEBI:73511
IUPAC Name hex-5-ynoic acid
SMILES C#CCCCC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.0160g/mL
Boiling Point 224.0°C to 225.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH≡C(CH2)3COOH
Refractive Index 1.4480 to 4.4500
Quantity 1 g
Specific Gravity 1.016
Formula Weight 112.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Hexynoic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Hexynoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.