missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hexyn-1-ol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
5-Hexyn-1-ol is used as starting material for the preparation of terminal alkynes and lactones containing terminal alykyne groups. It is also used in the synthesis of cinnoline-fused cyclic enediyne, llycopodium alkaloids, (+)-nankakurine A and (+)-nankakurine B and 7-benzoyloxy-3-(2-nitrophenylseleno)-1,5-cyclodecadiyne.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless to Yellow |
Density | 0.880 g/mL |
Boiling Point | 73°C to 75°C (15 mmHg) |
Flash Point | 70°C (158°F) |
Odor | Characteristic |
Linear Formula | HC≡C(CH2)3CH2OH |
Refractive Index | 1.4451 |
Quantity | 1 g |
Solubility Information | Slightly miscible with water. |
Formula Weight | 98.15 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'