missing translation for 'onlineSavingsMsg'
Learn More

5-Hexenenitrile, 95%

Product Code. p-142516 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
1mL
25mL
5mL
This item is not returnable. View return policy

Product Code. 10348903

Brand: Thermo Scientific Acros 259810010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5048-19-1 | C6H9N | 95.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5048-19-1
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.15
MDL Number MFCD00067056
InChI Key UNAQSRLBVVDYGP-UHFFFAOYSA-N
Synonym 5-hexenenitrile, 5-cyano-1-pentene, 4-pentenyl cyanide, hex-1-ene-6-nitrile, acmc-209kmu
PubChem CID 123222
IUPAC Name hex-5-enenitrile
SMILES C=CCCCC#N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 0.8200g/mL
Boiling Point 162.0°C to 163.0°C
Flash Point 40°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH2=CHCH2CH2CH2CN
Refractive Index 1.4250 to 1.4270
Quantity 1 mL
Specific Gravity 0.82
Solubility Information Solubility in water: soluble in hot water.
Formula Weight 95.15
Percent Purity 95%
Chemical Name or Material 5-Hexenenitrile, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Hexenenitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.