missing translation for 'onlineSavingsMsg'
Learn More

5-Hexen-2-one, 98%

Product Code. 11420697
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420697 5 g 5g
11430697 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420697 Supplier Thermo Scientific Alfa Aesar Supplier No. B22505.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-49-9 | C6H10O | 98.145 g/mol

Allyl Acetone is used as a nature identical flavor and fragrance. And it is also used to produce 2-methyl-hex-5-en-2-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl Acetone is used as a nature identical flavor and fragrance. And it is also used to produce 2-methyl-hex-5-en-2-ol.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-49-9
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00008793
InChI Key RNDVGJZUHCKENF-UHFFFAOYSA-N
Synonym 5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone
PubChem CID 7989
IUPAC Name hex-5-en-2-one
SMILES CC(=O)CCC=C

Specifications

Density 0.847
Boiling Point 127°C to 128°C
Flash Point 23°C (73°F)
Odor Like Ketone
Refractive Index 1.42
Quantity 5 g
UN Number UN1224
Beilstein 1699137
Solubility Information Soluble in water.
Formula Weight 98.15
Percent Purity ≥98%
Chemical Name or Material 5-Hexen-2-one
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.