missing translation for 'onlineSavingsMsg'
Learn More

5-Hexen-2-one, 98%

Product Code. 10070970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
20 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
20mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10070970 100 mL Glass bottle 100mL
10193381 5 mL Glass bottle 5mL
10764241 20 mL Glass bottle 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10070970 Supplier Thermo Scientific Acros Supplier No. 120781000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-49-9 | C6H10O | 98.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-49-9
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
MDL Number MFCD00008793
InChI Key RNDVGJZUHCKENF-UHFFFAOYSA-N
Synonym 5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone
PubChem CID 7989
IUPAC Name hex-5-en-2-one
SMILES CC(=O)CCC=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 0.8400g/mL
Boiling Point 128°C to 129°C
Flash Point 23°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2CH2COCH3
Refractive Index 1.4190 to 1.421
Quantity 100 mL
Beilstein 01, 734
Specific Gravity 0.84
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in organic solvents
Formula Weight 98.14
Percent Purity 98%
Chemical Name or Material 5-Hexen-2-one
Show More Show Less
compliance-icons
Product Identifier
  • 5-Hexen-2-one
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.