missing translation for 'onlineSavingsMsg'
Learn More

5-Hexen-1-ol, 98%

Product Code. 11422428
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11422428

Brand: Thermo Scientific Alfa Aesar A15766.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 821-41-0 | C6H12O | 100.161 g/mol

5-Hexen-1-ol is used in cyclization to a tetrahydropyran by phenylselenoetherification. It is also used as a building block in synthetic chemistry. Further, it is used to prepare 6-bromo-hex-1-ene by reaction with phosphorus tribromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hexen-1-ol is used in cyclization to a tetrahydropyran by phenylselenoetherification. It is also used as a building block in synthetic chemistry. Further, it is used to prepare 6-bromo-hex-1-ene by reaction with phosphorus tribromide.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-41-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00002981
InChI Key UIZVMOZAXAMASY-UHFFFAOYSA-N
Synonym 5-hexen-1-ol, 1-hexen-6-ol, 5-hexenol, unii-57pd1rf6g7, 5-hexene-1-ol, 5-hexanol, hex-1-en-6-ol, acmc-209pn9, ho ch2 4ch=ch2
PubChem CID 69963
IUPAC Name hex-5-en-1-ol
SMILES C=CCCCCO

Specifications

Melting Point <-20°C
Density 0.842
Boiling Point 152°C to 155°C
Flash Point 59°C (138°F)
Assay Percent Range 97%
Odor Characteristic
Refractive Index 1.436
Quantity 25 g
UN Number UN2924
Beilstein 1236458
Solubility Information Miscible with water.
Formula Weight 100.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Hexen-1-ol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.