missing translation for 'onlineSavingsMsg'
Learn More

5-(Hexadecanoylamino)fluorescein, TRC

Product Code. 30402888
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402888 1 g
30408069 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402888 Supplier Toronto Research Chemicals Supplier No. TRCH2909901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73024-80-3
Molecular Formula C36H43NO6
Molecular Weight (g/mol) 585.73
Synonym N-(3',6'-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9'-[9H]xanthen]-5-yl)hexadecanamide, H 110
IUPAC Name N-(3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-yl)hexadecanamide
SMILES CCCCCCCCCCCCCCCC(=O)Nc1ccc2c(c1)C(=O)OC23c4ccc(O)cc4Oc5cc(O)ccc35

Specifications

Quantity 1 g
InChI Formula InChI=1S/C36H43NO6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-34(40)37-25-16-19-29-28(22-25)35(41)43-36(29)30-20-17-26(38)23-32(30)42-33-24-27(39)18-21-31(33)36/h16-24,38-39H,2-15H2,1H3,(H,37,40)
Formula Weight 585.309
Chemical Name or Material 5-(Hexadecanoylamino)fluorescein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.