Learn More
Justrite™ 5 Gallon Drain Can
Product Code. 11924967
Change view
Click to view available options
Capacity (English):
757 L
Unit Size:
Each
Product Code. Capacity (English) Unit Size
11924967 757 L Each
1 options
Product Code. 11924967 Supplier Justrite™ Supplier No. 10906
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Justrite™ 5 Gallon Drain Can designed to easily and safely collect used solvents from parts when washing equipment.
  • Easy collection of used solvents
  • Wide-mouth funnel catches drainage
  • Built-in flame arrester
  • Wide-mouth funnel catches drainage with no spill, no mess
  • Removable funnel is plated steel
  • Built-in flame arrester keeps fire from contents
  • A 1.9 cm threaded plug allows safe disposal of waste liquids from the drain can into a permanent receptacle
  • Optional drain can cover fits on top of funel to keep out dirt
Specifications
Capacity (Metric) 5 Gallon
Capacity (English) 757 L
Color Yellow
Includes Plated steel funnel
Material Steel