missing translation for 'onlineSavingsMsg'
Learn More

5-Formylfuran-2-boronic acid, 95%

Product Code. 11384825
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384825 1 g 1g
11394825 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384825 Supplier Thermo Scientific Alfa Aesar Supplier No. L17920.03
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27329-70-0 | C5H5BO4 | 139.901 g/mol

As a bifunctional reagent used in the synthesis of π -extended heteroarylfuran systems. Reactant involved in Suzuki coupling for synthesis of stable dye-sensitized solar cells. Reactant involved in synthesis of biologically active molecules including, heteroarylation for the synthesis of HIF-1 inhibitors, disalicylic acid-furanyl derivatives to inhibit ephrin binding, HIV-1 integrase inhibitors, epidermal growth factor receptor inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As a bifunctional reagent used in the synthesis of π -extended heteroarylfuran systems. Reactant involved in Suzuki coupling for synthesis of stable dye-sensitized solar cells. Reactant involved in synthesis of biologically active molecules including, heteroarylation for the synthesis of HIF-1 inhibitors, disalicylic acid-furanyl derivatives to inhibit ephrin binding, HIV-1 integrase inhibitors, epidermal growth factor receptor inhibitors.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27329-70-0
Molecular Formula C5H5BO4
Molecular Weight (g/mol) 139.901
MDL Number MFCD01114696
InChI Key JUWYQISLQJRRNT-UHFFFAOYSA-N
Synonym 2-formylfuran-5-boronic acid, 5-formylfuran-2-yl boronic acid, 5-formyl-2-furanboronic acid, 5-formylfuran-2-boronic acid, 5-formyl-2-furylboronic acid, 5-formyl-2-furanylboronic acid, 2-formylfuran-5-boronicacid, 5-formyl-2-furyl boronic acid, 5-formyl-2-furanyl boronic acid, boronic acid, 5-formyl-2-furanyl
PubChem CID 2734355
IUPAC Name (5-formylfuran-2-yl)boronic acid
SMILES B(C1=CC=C(O1)C=O)(O)O

Specifications

Melting Point ∽140°C (decomposition)
Quantity 1 g
Beilstein 1637050
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 139.9
Percent Purity 95%
Chemical Name or Material 5-Formylfuran-2-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.