missing translation for 'onlineSavingsMsg'
Learn More

5-Formylcytosines Antibody Pair, Biotin, Novus Biologicals™

Product Code. 18672414 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18672414 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18672414 Supplier Novus Biologicals Supplier No. NBP323469

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

5-Formylcytosines Monoclonal antibody specifically detects 5-Formylcytosines in Human samples. It is validated for ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5-Formylcytosines
Applications ELISA
Classification Monoclonal
Conjugate Biotin
Dilution ELISA, ELISA Capture (Matched Antibody Pair) 50 - 200ng/well, ELISA Detection (Matched Antibody Pair) 0.1 - 0.5ug/ml
Host Species Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.