missing translation for 'onlineSavingsMsg'
Learn More

5-Formyl-heptanoic Acid, TRC

Product Code. 30401682
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401682 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401682 Supplier Toronto Research Chemicals Supplier No. TRCF700513250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Formyl-heptanoic Acid
CAS 1379186-31-8
Molecular Formula C8H14O3
Quantity 250 mg
Synonym 5-Formylheptanoic Acid
InChI Formula InChI=1S/C8H14O3/c1-2-7(6-9)4-3-5-8(10)11/h6-7H,2-5H2,1H3,(H,10,11)
SMILES CCC(CCCC(=O)O)C=O
IUPAC Name 5-formylheptanoic acid
Molecular Weight (g/mol) 158.19
Formula Weight 158.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.