missing translation for 'onlineSavingsMsg'
Learn More

5-Formyl-2'-O-methylcytidine, TRC

Product Code. 30396640
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396640 5 mg 5mg
30392837 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396640 Supplier Toronto Research Chemicals Supplier No. TRCF7016605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Formyl-2’-O-methylcytidine
CAS 176858-79-0
Molecular Formula C11 H15 N3 O6
Quantity 5 mg
Synonym 4-Amino-1-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-2-oxo-1,2-dihydropyrimidine-5-carbaldehyde
InChI Formula InChI=1S/C11H15N3O6/c1-19-8-7(17)6(4-16)20-10(8)14-2-5(3-15)9(12)13-11(14)18/h2-3,6-8,10,16-17H,4H2,1H3,(H2,12,13,18)/t6-,7-,8-,10-/m1/s1
SMILES CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N2C=C(C=O)C(=NC2=O)N
IUPAC Name 4-amino-1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxy-tetrahydrofuran-2-yl]-2-oxo-pyrimidine-5-carbaldehyde
Molecular Weight (g/mol) 285.253
Formula Weight 285.0961
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.