missing translation for 'onlineSavingsMsg'
Learn More

Melford 5-FOA

Product Code. 30018270
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
10 g
25 g
Unit Size:
100mg
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018270 5 g 5g
30018267 1 g 1g
30018269 25 g 25g
30018268 10 g 10g
30018266 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30018270 Supplier Melford Supplier No. F105015.0
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Reagent for selection of Orotidine-5'-phosphate decarboxylase mutants of Saccharomyces cerevisiae.

Reagent for selection of Orotidine-5'-phosphate decarboxylase mutants of Saccharomyces cerevisiae.

Specifications

CAS 703-95-7
Molecular Formula C5H3FN2O4
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.