missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford 5-FOA
Click to view available options
Quantity:
100 mg
1 g
5 g
10 g
25 g
Unit Size:
100mg
10g
1g
25g
5g
Description
Reagent for selection of Orotidine-5'-phosphate decarboxylase mutants of Saccharomyces cerevisiae.
Reagent for selection of Orotidine-5'-phosphate decarboxylase mutants of Saccharomyces cerevisiae.
Specifications
Specifications
| CAS | 703-95-7 |
| Molecular Formula | C5H3FN2O4 |
| Quantity | 10 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction