Learn More
5-Fluorouridine, 97%
Description
5-Fluorouridine exhibits anticancer activity. It is often used in chemical and biochemical comparison studies with fluorouracil and thymine analogs. It is an active metabolite of Doxifluridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
5-Fluorouridine exhibits anticancer activity. It is often used in chemical and biochemical comparison studies with fluorouracil and thymine analogs. It is an active metabolite of Doxifluridine.
Solubility
Soluble in Methanol (Slightly), Water (Slightly).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Store at 4°C.
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Beilstein | 33662 |
| Solubility Information | Soluble in Methanol (Slightly),Water (Slightly). |
| Formula Weight | 262.19 |
| Percent Purity | 97% |
| Physical Form | Powder |
| Chemical Name or Material | 5-Fluorouridine |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.