missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluorouridine 5'-Triphosphate Ammonium Salt (>80%), TRC
Specifications
Specifications
| Chemical Name or Material | 5-Fluorouridine 5'-Triphosphate Ammonium Salt |
| Analyte or Component Names | 5-Fluorouridine 5'-Triphosphate Ammonium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9H14FN2O15P3 • x(NH3) |
| Synonym | ((2R,3S,4R,5R)-5-(5-fluoro-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate ammonium salt |
| InChI Formula | InChI=1S/C9H14FN2O15P3/c10-3-1-12(9(16)11-7(3)15)8-6(14)5(13)4(25-8)2-24-29(20,21)27-30(22,23)26-28(17,18)19/h1,4-6,8,13-14H,2H2,(H,20,21)(H,22,23)(H,11,15,16)(H2,17,18,19)/t4-,5-,6-,8-/m1/s1 |
| Solution Type | Neat |
| SMILES | O=C1C(F)=CN([C@H]2[C@H](O)[C@H](O)[C@@H](COP(O)(OP(O)(OP(O)(O)=O)=O)=O)O2)C(N1)=O |
| IUPAC Name | ((2R,3S,4R,5R)-5-(5-fluoro-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?