missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorouracil N-Beta-D-Glucuronide Methyl Ester, 2,3,4-Triacetate, TRC

Product Code. 30399294
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399294 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399294 Supplier Toronto Research Chemicals Supplier No. TRCF59601550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Fluorouracil N-Beta-D-Glucuronide Methyl Ester, 2,3,4-Triacetate
CAS 77476-81-4
Molecular Formula C17H19FN2O11
Quantity 50 mg
Synonym 1-Deoxy-1-(5-fluoro-3,4-dihydro-2,4-dioxo-1(2H)-pyrimidinyl)-β-D-glucopyranuronic Acid Methyl Ester, 2,3,4-Triacetate
InChI Formula InChI=1S/C17H19FN2O11/c1-6(21)28-10-11(29-7(2)22)13(16(25)27-4)31-15(12(10)30-8(3)23)20-5-9(18)14(24)19-17(20)26/h5,10-13,15H,1-4H3,(H,19,24,26)/t10-,11-,12+,13-,15+/m0/s1
SMILES COC(=O)[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C)N2C=C(F)C(=O)NC2=O
Molecular Weight (g/mol) 446.34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.