missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorouracil, MedChemExpress

Product Code. p-200096825
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16431999

Brand: Medchem Express HY90006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 5-Fluorouracil (5-FU) is an analogue of uracil and a potent antitumor agent. 5-Fluorouracil affects pyrimidine synthesis by inhibiting thymidylate synthetase thus depleting intracellular dTTP pools. 5-Fluorouracil induces apoptosis and can be used as a chemical sensitizer. 5-Fluorouracil also inhibits HIV.

5-Fluorouracil (5-FU) is an analogue of uracil and a potent antitumor agent. 5-Fluorouracil affects pyrimidine synthesis by inhibiting thymidylate synthetase thus depleting intracellular dTTP pools. 5-Fluorouracil induces apoptosis and can be used as a chemical sensitizer. 5-Fluorouracil also inhibits HIV.

Specifications

Chemical Name or Material 5-Fluorouracil
Physical Form Solid
CAS 51-21-8
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C4H3FN2O2
For Use With (Application) Cancer-programmed cell death
Synonym 5-FU
Solubility Information DMSO : 50 mg/mL (384.38 mM; Need ultrasonic) ∣H2O : 20 mg/mL (153.75 mM; Need ultrasonic)
SMILES O=C(N1)NC=C(F)C1=O
Molecular Weight (g/mol) 130.08
Formula Weight 130.08
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.