missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorouracil, MedChemExpress

Product Code. 16460698
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460698 5 g 5g
16431999 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460698 Supplier Medchem Express Supplier No. HY90006
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5-Fluorouracil (5-FU) is an analogue of uracil and a potent antitumor agent. 5-Fluorouracil affects pyrimidine synthesis by inhibiting thymidylate synthetase thus depleting intracellular dTTP pools. 5-Fluorouracil induces apoptosis and can be used as a chemical sensitizer. 5-Fluorouracil also inhibits HIV.

5-Fluorouracil (5-FU) is an analogue of uracil and a potent antitumor agent. 5-Fluorouracil affects pyrimidine synthesis by inhibiting thymidylate synthetase thus depleting intracellular dTTP pools. 5-Fluorouracil induces apoptosis and can be used as a chemical sensitizer. 5-Fluorouracil also inhibits HIV.

Specifications

Chemical Name or Material 5-Fluorouracil
Physical Form Solid
CAS 51-21-8
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C4H3FN2O2
For Use With (Application) Cancer-programmed cell death
Synonym 5-FU
Solubility Information DMSO : 50 mg/mL (384.38 mM; Need ultrasonic) ∣H2O : 20 mg/mL (153.75 mM; Need ultrasonic)
SMILES O=C(N1)NC=C(F)C1=O
Molecular Weight (g/mol) 130.08
Formula Weight 130.08
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.