missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorouracil, >99%, MP Biomedicals™

Product Code. 11498070 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498070 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11498070 Supplier MP Biomedicals Supplier No. 0210172201
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 51-21-8
Molecular Formula C4H3FN2O2
MDL Number MFCD00006018
Quantity 1 g
Synonym 5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, 5-fluoropyrimidine-2,4 1h,3h-dione
InChI Key GHASVSINZRGABV-UHFFFAOYSA-N
SMILES FC1=CNC(=O)NC1=O
IUPAC Name 5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Weight (g/mol) 130.08
PubChem CID 3385
ChEBI CHEBI:46345
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.