missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-Fluorouracil, 99%

Product Code. 10697552 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10697552 5 g Glass bottle 5g
10348170 1 g Glass bottle 1g
11906531 25 g Glass bottle 25g
15159263 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10697552 Supplier Thermo Scientific Chemicals Supplier No. 228440050
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

5-Fluorouracil, CAS # 51-21-8, is an analogue base composed of uracil with a position 5 hydrogen fluorine substitution. | CAS: 51-21-8 | C4H3FN2O2 | 130.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 5-Fluorouracil is an antimetabolite and antineoplastic that prevents DNA synthesis by blocking conversion of deoxyuridylic acid

• 5-Fluorouracil can replace uracil in RNA and prevent protein synthesis and cell growth

Applications

• 5-Fluorouracil can be used for laboratory research on tumor growth and immunosuppression

• 5-Fluorouracil may also be appropriate for laboratory studies of neurotoxicity, cell lysis, and DNA damage

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-21-8
Molecular Formula C4H3FN2O2
Molecular Weight (g/mol) 130.08
MDL Number MFCD00006018
InChI Key GHASVSINZRGABV-UHFFFAOYSA-N
Synonym 5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, kecimeton
PubChem CID 3385
ChEBI CHEBI:46345
IUPAC Name 5-fluoro-1H-pyrimidine-2,4-dione
SMILES FC1=CNC(=O)NC1=O

Specifications

Melting Point 278.0°C to 282.0°C
Color White
Boiling Point 190.0°C to 200.0°C (0.1 mmHg)
Loss on Drying 0.5% (80°C, 4 hrs) (in vacuum) max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 24,IV,1229
Merck Index 15,4213
Solubility Information Solubility in water: 12.2g/L (20°C). Other solubilities: 2.9g/L (20°C) in ethanol,readily soluble in aqueous alkali solutions,soluble in dmso
Formula Weight 130.08
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorouracil
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Fluorouracil
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H361d-Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.