missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroorotic Acid Monohydrate (Ultra Pure), TRC

Product Code. 17964485
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17964485 5 g 5g
17932935 25 g 25g
17902065 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17964485 Supplier Toronto Research Chemicals Supplier No. TRCF5950005G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220141-70-8
Molecular Formula C5H5FN2O5
Molecular Weight (g/mol) 192.1
SMILES OC(=O)C1=C(F)C(=O)NC(=O)N1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H3FN2O4.H2O/c6-1-2(4(10)11)7-5(12)8-3(1)9;/h(H,10,11)(H2,7,8,9,12);1H2
Percent Purity >95%
Chemical Name or Material 5-Fluoroorotic Acid, Monohydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.