missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroorotic acid monohydrate, 98%, Thermo Scientific™

Product Code. 11389709
Change view
Click to view available options
Quantity:
50 mg
1 g
Unit Size:
1g
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389709 50 mg 50mg
11399709 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389709 Supplier Thermo Scientific Alfa Aesar Supplier No. B21290.MF

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Fluoroorotic Acid, Monohydrate (5-FOA) has become a valuable tool for research in the field of molecular genetics and has been employed in the selection of resistant strains of Saccharomyces cerevisiae that possess a mutant URA3 gene which renders them orotidine-5í-phosphate decarboxylation.

Solubility
Soluble in water, dimethyl sulfoxide, 50mg in 1ml (1:1 ammonium hydroxide: water) with gentle heating.

Notes
Store in freezer (-20°C). Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220141-70-8
Molecular Formula C5H5FN2O5
Molecular Weight (g/mol) 192.10
MDL Number MFCD00150658
InChI Key LODRRYMGPWQCTR-UHFFFAOYSA-N
Synonym 5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, 5-fluoroorotic acid monohydrate, 5-fluoroorotic acid hydrate, 5-fluoroorotic acid,monohydrate, 5-fluoro-1,2,3,6-tetrahydro-2,6-dioxopyrimidine-4-carboxylic acid hydrate, 5-fluoroortic acid monohydrate, 5-fluorouracil-4-carboxylic acid hydrate, 5-fluoroorotic acid n-hydrate, 5-fluororotic acid monohydrate, 5-fluoro-orotic acid monohydrate
PubChem CID 16212749
SMILES O.OC(=O)C1=C(F)C(=O)NC(=O)N1

Specifications

Melting Point ∼255°C (decomposition)
Quantity 50 mg
Beilstein 181933
Merck Index 14,4176
Solubility Information Soluble in water,dimethyl sulfoxide,50mg in 1ml (1:1 ammonium hydroxide: water) with gentle heating.
IUPAC Name 5-fluoro-2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
Formula Weight 192.11 (174.09 Anhydrous)
Percent Purity 98%
Chemical Name or Material 5-Fluoroorotic acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.