missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroorotic Acid Hydrate, TRC

Product Code. 30401747
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401747 250 mg
30401947 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401747 Supplier Toronto Research Chemicals Supplier No. TRCF595078250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207291-81-4
Molecular Formula C5H3FN2O4 . xH2O
Molecular Weight (g/mol) 192.1
SMILES OC(=O)C1=C(F)C(=O)NC(=O)N1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H3FN2O4.H2O/c6-1-2(4(10)11)7-5(12)8-3(1)9;/h(H,10,11)(H2,7,8,9,12);1H2
Formula Weight 192.0182
Chemical Name or Material 5-Fluoroorotic Acid Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.