missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindole-3-carboxaldehyde, 99%, Thermo Scientific™

Product Code. 15214754 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15214754 5 g 5g
15204754 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15214754 Supplier Thermo Scientific Acros Supplier No. 443350050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2338-71-8
Molecular Formula C9H6FNO
Molecular Weight (g/mol) 163.15
MDL Number MFCD00022719
InChI Key YUAJKGBLPVLADK-UHFFFAOYSA-N
Synonym 5-fluoroindole-3-carboxaldehyde, 5-fluoro-1h-indole-3-carboxaldehyde, 5-fluoroindole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 5-fluoro, 3-formyl-5-fluoroindole, pubchem7267, zlchem 1197, 5-fluoro-3-formylindole, 5-fluoroindole-3-aldehyde, acmc-209g2v
PubChem CID 259089
IUPAC Name 5-fluoro-1H-indole-3-carbaldehyde
SMILES FC1=CC=C2NC=C(C=O)C2=C1

Specifications

Melting Point 170°C to 172°C
Assay Percent Range 98.5% min. (HPLC)
Quantity 5 g
Formula Weight 163.15
Percent Purity 99%
Chemical Name or Material 5-Fluoroindole-3-carboxaldehyde
compliance-icons
Product Identifier
  • 5-Fluoroindole-3-carboxaldehyde, 99%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.