missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorocytosine, 99+%

Product Code. 10742354 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10742354 5 g Glass bottle 5g
10125793 250 mg Glass bottle 250mg
10204381 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10742354 Supplier Thermo Scientific Acros Supplier No. 258340050
  • ONLINE EXCLUSIVE PRICE
    £520.00
    £434.65 / 5g
    Save £85.35
    16% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2022-85-7 | C4H4FN3O | 129.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2022-85-7
Molecular Formula C4H4FN3O
Molecular Weight (g/mol) 129.09
MDL Number MFCD00006035,MFCD00179326,MFCD03547958
InChI Key XRECTZIEBJDKEO-UHFFFAOYSA-N
Synonym 5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
PubChem CID 3366
ChEBI CHEBI:5100
IUPAC Name 6-amino-5-fluoro-1H-pyrimidin-2-one
SMILES NC1=C(F)C=NC(=O)N1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 296.0°C
Color White
Loss on Drying 1.0% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Beilstein 25, 3688
Merck Index 15, 4156
Solubility Information Solubility in water: 15g/L (25°C)
Formula Weight 129.09
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorocytosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.