missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluorobenzoxazole, 97%, Thermo Scientific™
£67.10 - £204.00
Chemical Identifiers
CAS | 221347-71-3 |
---|---|
Molecular Formula | C7H4FNO |
Molecular Weight (g/mol) | 137.11 |
InChI Key | ZRMPAEOUOPNNPZ-UHFFFAOYSA-N |
Synonym | 5-fluorobenzoxazole, 5-fluorobenzo d oxazole, benzoxazole, 5-fluoro, benzoxazole,5-fluoro, pubchem21888, 5-fluoranyl-1,3-benzoxazole, benzoxazole, 5-fluoro-9ci |
PubChem CID | 19937167 |
IUPAC Name | 5-fluoro-1,3-benzoxazole |
SMILES | C1=CC2=C(C=C1F)N=CO2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15355028
|
Thermo Scientific Acros
455612500 |
250 mg |
£67.10
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15345028
|
Thermo Scientific Acros
455610010 |
1 g |
£204.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
221347-71-3 | |
137.11 | |
5-fluorobenzoxazole, 5-fluorobenzo d oxazole, benzoxazole, 5-fluoro, benzoxazole,5-fluoro, pubchem21888, 5-fluoranyl-1,3-benzoxazole, benzoxazole, 5-fluoro-9ci | |
5-fluoro-1,3-benzoxazole |
C7H4FNO | |
ZRMPAEOUOPNNPZ-UHFFFAOYSA-N | |
19937167 | |
C1=CC2=C(C=C1F)N=CO2 |
Specifications
221347-71-3 | |
96% min. (GC) | |
5-fluorobenzoxazole, 5-fluorobenzo d oxazole, benzoxazole, 5-fluoro, benzoxazole,5-fluoro, pubchem21888, 5-fluoranyl-1,3-benzoxazole, benzoxazole, 5-fluoro-9ci | |
C1=CC2=C(C=C1F)N=CO2 | |
137.11 | |
137.11 | |
5-Fluorobenzoxazole |
Authentic | |
C7H4FNO | |
ZRMPAEOUOPNNPZ-UHFFFAOYSA-N | |
5-fluoro-1,3-benzoxazole | |
19937167 | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physicia
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 5-Fluorobenzoxazole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only