missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroanthranilic Acid, TRC

Product Code. 30401584
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401584 250 mg
30402975 1 g
30390431 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401584 Supplier Toronto Research Chemicals Supplier No. TRCF587750250MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-08-2
Molecular Formula C7 H6 F N O2
Molecular Weight (g/mol) 155.13
IUPAC Name 2-amino-5-fluorobenzoic acid
SMILES Nc1ccc(F)cc1C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H6FNO2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3H,9H2,(H,10,11)
Formula Weight 155.0383
Chemical Name or Material 2-Amino-5-fluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.