missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroanthranilic Acid, TRC

Product Code. 30390431
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390431 5 g
30401584 250 mg
30402975 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390431 Supplier Toronto Research Chemicals Supplier No. TRCF5877505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-08-2
Molecular Formula C7 H6 F N O2
Molecular Weight (g/mol) 155.13
IUPAC Name 2-amino-5-fluorobenzoic acid
SMILES Nc1ccc(F)cc1C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H6FNO2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3H,9H2,(H,10,11)
Formula Weight 155.0383
Chemical Name or Material 2-Amino-5-fluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.