missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro Uracil, TRC

Product Code. 30008773
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008773 100 g 100g
30005145 10 g 10g
18020850 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008773 Supplier Toronto Research Chemicals Supplier No. TRCF596000100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-21-8
Molecular Formula C4 H3 F N2 O2
Molecular Weight (g/mol) 130.08
Synonym Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, Flucytosine Imp. A (EP), 2,4(1H,3H)-Pyrimidinedione, 5-fluoro-, Uracil, 5-fluoro- (8CI), 5-Fluoro-2,4(1H,3H)-pyrimidinedione, 2,4-Dihydroxy-5-fluoropyrimidine, 2,4-Dioxo-5-fluoropyrimidine, 5-FU, 5-Fluoracyl, 5-Fluoro-1H-pyrimidine-2,4-dione, 5-Fluoro-2,4-dihydroxypyrimidine, 5-Fluoro-2,4-pyrimidinedione, 5-Fluoropyrimidine-2,4-diol, 5-Fluorouracil, 5-Oncouracil, Adrucil, Arumel, Carac, Carzonal, DBL, Efudex, Efudix, Efurix, FU, Fluoroblastin, Fluoroplex, Fluorouracile Accord, Flurablastin, Fluracedyl, Fluracil, Fluracilum, Fluri, Fluril, Ftoruracil, Kecimeton, NSC 19893, Phthoruracil, Phtoruracil, Queroplex, Ro 2-9757, Sigma 03738, Timazin, U 8953, Ulup, Fluorouracil
IUPAC Name 5-fluoro-1H-pyrimidine-2,4-dione
SMILES FC1=CNC(=O)NC1=O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)
Formula Weight 130.0179
Percent Purity >95%
Chemical Name or Material 5-Fluorouracil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.