missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro Isatin, TRC

Product Code. 30398323
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398323 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398323 Supplier Toronto Research Chemicals Supplier No. TRCF59200010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Fluoroisatin
CAS 443-69-6
Molecular Formula C8 H4 F N O2
Quantity 10 g
Synonym 1H-Indole-2,3-dione, 5-fluoro-, Indole-2,3-dione, 5-fluoro- (7CI,8CI), Isatin, 5-fluoro- (6CI), 5-Fluoro-1H-indole-2,3-dione, 5-Fluoro-2,3-indoledione, 5-Fluoroindoline-2,3-dione, 5-Fluoroisatin, 5-Fluoroisatine, NSC 39161
InChI Formula InChI=1S/C8H4FNO2/c9-4-1-2-6-5(3-4)7(11)8(12)10-6/h1-3H,(H,10,11,12)
SMILES Fc1ccc2NC(=O)C(=O)c2c1
IUPAC Name 5-fluoro-1H-indole-2,3-dione
Molecular Weight (g/mol) 165.12
Formula Weight 165.0226
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.