missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro ent-Lamivudine Acid D-Menthol Ester, TRC

Product Code. 30390058
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390058 50 mg
30392434 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390058 Supplier Toronto Research Chemicals Supplier No. TRCF59209050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 764659-79-2
Molecular Formula C18H26FN3O4S
Molecular Weight (g/mol) 399.48
Synonym (2S,5R)-5-(4-Amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylic Acid (1S,2R,5S)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester
IUPAC Name [(1S,2R,5S)-5-methyl-2-propan-2-ylcyclohexyl] (2S,5R)-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-1,3-oxathiolane-2-carboxylate
SMILES CC(C)[C@H]1CC[C@H](C)C[C@@H]1OC(=O)[C@H]2O[C@H](CS2)N3C=C(F)C(=NC3=O)N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H26FN3O4S/c1-9(2)11-5-4-10(3)6-13(11)25-16(23)17-26-14(8-27-17)22-7-12(19)15(20)21-18(22)24/h7,9-11,13-14,17H,4-6,8H2,1-3H3,(H2,20,21,24)/t10-,11+,13-,14+,17-/m0/s1
Formula Weight 399.1628
Chemical Name or Material 5-Fluoro ent-Lamivudine Acid D-Menthol Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.