missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-D,L-lysine Hydrochloride (90%), TRC

Product Code. 30404314
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404314 0.5 mg 0.5mg
30404165 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30404314 Supplier Toronto Research Chemicals Supplier No. TRCF5921000.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58960-25-1
Molecular Formula C6H13FN2O2 . HCl
Molecular Weight (g/mol) 200.64
Synonym 5-Fluoro-lysine Hydrochloride, 5-Fluoro-lysine Monohydrochloride
SMILES Cl.NCC(F)CCC(N)C(=O)O

Specifications

InChI Formula InChI=1S/C6H13FN2O2.ClH/c7-4(3-8)1-2-5(9)6(10)11;/h4-5H,1-3,8-9H2,(H,10,11);1H
Quantity 0.5 mg
Shipping Condition Room Temperature
Chemical Name or Material 5-Fluoro-D,L-lysine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.