missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro Cytosine, TRC

Product Code. 30132659
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132659 500 mg 500mg
30133193 1 g 1g
17941635 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132659 Supplier Toronto Research Chemicals Supplier No. TRCF589000500MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2022-85-7
Molecular Formula C4 H4 F N3 O
Molecular Weight (g/mol) 129.09
Synonym Flucytosine, 2(1H)-Pyrimidinone, 6-amino-5-fluoro-, 2(1H)-Pyrimidinone, 4-amino-5-fluoro- (9CI), Cytosine, 5-fluoro- (6CI,7CI,8CI), 6-Amino-5-fluoro-2(1H)-pyrimidinone, 4-Amino-5-fluoropyrimidin-2(1H)-one, 4-Amino-5-fluoropyrimidin-2-ol, 5-Fluorocytosin, 5-Fluorocytosine, 6-Amino-2-oxo-5-fluoropyrimidine, Alcobon, Ancobon, Ancotil, Ancotyl, Flucytosin, Fluocytosine, Fluorocytosine, NSC 103805, Ro 2-9915
IUPAC Name 4-amino-5-fluoro-1H-pyrimidin-2-one
SMILES NC1=NC(=O)NC=C1F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H4FN3O/c5-2-1-7-4(9)8-3(2)6/h1H,(H3,6,7,8,9)
Formula Weight 129.0338
Chemical Name or Material 5-Fluorocytosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.