missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride, 96%

Product Code. p-7031828
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448285 1 g 1g
15458285 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448285 Supplier Thermo Scientific Alfa Aesar Supplier No. H33044.03
Add to basket
This item is not returnable. View return policy

CAS: 84163-16-6 | C12H14ClFN2O | 256.705 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84163-16-6
Molecular Formula C12H14ClFN2O
Molecular Weight (g/mol) 256.705
MDL Number MFCD06200879
InChI Key HSOJFILHZZEWJT-UHFFFAOYSA-N
Synonym 5-fluoro-3-piperidin-4-yl benzo d isoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride, 5-fluoro-3-4-piperidinyl-1,2-benzisoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl-1,2-benzoxazole hydrochloride, 5-fluoro-3-piperidin-4-yl benzo d isoxazole hcl, 4-5-fluoro-1,2-benzisoxazol-3-yl piperidine hydrochloride, 1,2-benzisoxazole,5-fluoro-3-4-piperidinyl-, hydrochloride, pubchem11469, 5-fluoro-3-piperidin-4-yl benzisoxazole hcl salt, 5-fluoro-3-4-piperidyl benzo d isoxazole, chloride
PubChem CID 2794754
IUPAC Name 5-fluoro-3-piperidin-4-yl-1,2-benzoxazole;hydrochloride
SMILES C1CNCCC1C2=NOC3=C2C=C(C=C3)F.Cl

Specifications

Melting Point ∽298°C (decomposition)
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 256.71
Percent Purity 96%
Chemical Name or Material 5-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.