missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-(trifluoromethyl)phenol, 97%, Thermo Scientific Chemicals

Product Code. 15485157
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15485157 5 g 5g
15475157 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485157 Supplier Thermo Scientific Alfa Aesar Supplier No. H32445.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

As raw material in organic synthesis. Utilized in the dye industry. As pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As raw material in organic synthesis. Utilized in the dye industry. As pharmaceutical intermediate.

Solubility
Sparingly Soluble (0.40 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 243459-91-8
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.102
MDL Number MFCD00236302
InChI Key QWLZSSYHAJSEHU-UHFFFAOYSA-N
PubChem CID 2774803
IUPAC Name 5-fluoro-2-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1F)O)C(F)(F)F

Specifications

Melting Point 48°C to 52°C
Quantity 5 g
UN Number UN3261
Synonym 5-fluoro-2-trifluoromethyl phenol, 5-fluoro-2-trifluoromethyl-phenol, 4-fluoro-2-hydroxybenzotrifluoride, pubchem2171, intermediates-zcf02657, phenol,5-fluoro-2-trifluoromethyl, phenol, 5-fluoro-2-trifluoromethyl
Solubility Information Sparingly Soluble (0.40g/L) (25°C).
Formula Weight 180.1
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.