missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™

Product Code. 11366809
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11366809 1 g 1g
11376809 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366809 Supplier Thermo Scientific Alfa Aesar Supplier No. H26751.03
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as pharmaceutical intermediate.

Solubility
Soluble in organic solvents

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 231291-14-8
Molecular Formula C8H7F4N
Molecular Weight (g/mol) 193.145
MDL Number MFCD00236288
InChI Key ZRDWPCJUMDGSCG-UHFFFAOYSA-N
Synonym 5-fluoro-2-trifluoromethyl benzylamine, 5-fluoro-2-trifluoromethyl phenyl methanamine, 5-fluoro-2-trifluoromethyl phenyl methylamine, 1-5-fluoro-2-trifluoromethyl phenyl methanamine, acmc-1cqk1, ksc494q0r, 2-trifluoromethyl-5-fluorobenzylamine, 5-fluoro-2-trifluoromethylbenzylamine, 5-fluoro-2-trifluoromethyl phenyl methamine
PubChem CID 2737568
IUPAC Name [5-fluoro-2-(trifluoromethyl)phenyl]methanamine
SMILES C1=CC(=C(C=C1F)CN)C(F)(F)F

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Soluble in organic solvents
Formula Weight 193.15
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.